Skip to Content
Merck
All Photos(1)

Documents

B3051

Millipore

Brucella broth base

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

7.0±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
sample preparation

microbiology

suitability

selective for Brucella spp.
selective for Campylobacter spp.

Related Categories

Application

Recommended for enrichment, isolation and cultivation of Brucella species.

Components

Components (g/L):
Casein Enzymic Hydrolysate 10.00
Peptic Digest of Animal Tissue 10.00
Yeast Extract 2.00
Dextrose 1.00
Sodium Chloride 5.00
Sodium bisulphite 0.10

Preparation Note

Suspend 14.05 g of Brucella Broth Base in 500 ml of distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool to 45-50°C and aseptically add sterile 5% v/v heat inactivated (H1138) horse serum. Add polymyxin B (2,500 IU, Cat. No. 81334), bacitracin (12,500 IU, Cat. No. 11702), nystatin (50,000 IU, Cat. No. 74721), cycloheximide (50mg, Cat. No. 01810), nalidixic acid (2.5mg, Cat. No. 70162) and vancomycin (10mg, Cat. No. 94747) to make the medium selective. Mix well before pouring into sterile petri plates or tubes.
For isolation of the Campylobacter species add the rehydrated contents of one vial of Campylobacter Growth Supplement (Cat. No. C2847) and 5-7% defibrinated sheep blood to 500 ml of medium base. 500 ml of medium base. The Campylobacter Growth Supplement can also be made by mixing 125 mg sodium pyruvate (Cat. No. 15990), 125 mg sodium metabisulfite (Cat. No. 71928 )and 125 mg ferrous sulfate (Cat. No. 44970) in 2 ml of distilled water and filtering the solution through a sterile 0.4 mm filter to sterilize.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mohsen Tabasi et al.
Comparative immunology, microbiology and infectious diseases, 63, 87-93 (2019-04-10)
Brucellosis is the most common zoonotic diseases worldwide. The aim of this study was to develop and evaluate the diagnostic performance of an indirect-ELISA (I-ELISA) method based on whole cell Brucella abortus S99 lysates for detection of IgM anti-Brucella antibodies
Carla Palacios-Gorba et al.
Food & function, 11(5), 4525-4534 (2020-05-13)
Currently, Helicobacter pylori is the unique biological carcinogenic agent. The search for antimicrobial alternatives to antibiotics against this pathogen has been categorized as a priority due to the drastic failure associated with current applied antibiotic therapy. The present study assessed
A selective medium for the isolation of brucellae from contaminated materials.
C D KUZDAS et al.
Journal of bacteriology, 66(4), 502-504 (1953-10-01)
M Boyle et al.
Applied and environmental microbiology, 74(10), 2997-3001 (2008-03-25)
Batch solar disinfection (SODIS) inactivation kinetics are reported for suspensions in water of Campylobacter jejuni, Yersinia enterocolitica, enteropathogenic Escherichia coli, Staphylococcus epidermidis, and endospores of Bacillus subtilis, exposed to strong natural sunlight in Spain and Bolivia. The exposure time required
A preliminary report on a selective medium for the culture of Brucella, including fastidious types.
L M JONES et al.
Bulletin of the World Health Organization, 19(1), 200-203 (1958-01-01)

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Related Content

Step-by-step protocol for generating apical-out human gut organoids for microbiome, ADME/Tox, viral and gastrointestinal related disease research. See the complete organoid culture protocol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service