Skip to Content
Merck
All Photos(1)

Key Documents

1.00070

Millipore

mCCD (Modified charcoal cefoperazone deoxycholate) agar (base)

acc. ISO 10272 GranuCult®

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€302.00

€302.00


In StockDetails


Request more information

Select a Size

Change View
500 G
€302.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€302.00


In StockDetails


Request more information

Agency

ISO

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.2-7.6 (25 °C, 44.5 g/L in H2O)

solubility

45.5 g/L

application(s)

food and beverages
microbiology
pathogen testing

storage temp.

15-25°C

suitability

Campylobacter spp.

General description

GranuCult is our new brand name for our existing granulated dehydrated culture media which apply to new EN ISO 11133:2014 and various other ISO standards and FDA-BAM / USDA-FSIS methods for testing of food and water samples. All these media are currently being updated to be fully compliant to the latest version of these standards and regulations with respect to composition and quality control and will be available shortly.

Analysis Note

Appearance (colour): black
pH-value (25 °C): 7.2 - 7.6
Typical composition (g/litre): Peptones 20.0; Casein hydrolysate 30.; Activated charcoal 4.0; Sodium deoxycholate 1.0; Sodium pyruvate 0.25; Ferrous sulfate 0.25; Agar-agar 12.0.
Growth promotion test in accordance with ISO 11133.
Base tested with supplement Art. 1.00071.
Growth (Campylobacter coli ATCC 43478 (WDCM 00004)): good
Growth (Campylobacter jejuni ATCC 33291 (WDCM 00005)): good
Growth (Campylobacter jejuni ATCC 29428 (WDCM 00156)): good
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): no growth
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): inhibited growth
Growth (Escherichia coli ATCC 8739 (WDCM 00012)): inhibited growth
Incubation: 44 hrs. 41.5 °C with Anaerocult C. E.coliand Staph.aur. aerobic.
Strains tested with three-loop-streak.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service