94791
SnapIT Ampule Opener
Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
All Photos(2)
About This Item
Recommended Products
General description
SnapITs are easy-to-use, multi-use ampule openers that avoid the possibility of injury sometimes seen when ampoules are opened by hand.
1. INSERT the ampoule lid into your multi-use SnapIT
2. SNAP open the ampoule
3. EJECT the ampoule lid directly into a sharps container
1. INSERT the ampoule lid into your multi-use SnapIT
2. SNAP open the ampoule
3. EJECT the ampoule lid directly into a sharps container
Choose from one of the most recent versions:
Certificates of Analysis (COA)