Documents
Z150770
Glass-tubing cutter
About This Item
UNSPSC Code:
41122406
NACRES:
NB.35
Related Categories
General description
Spring-loaded to stay open until used. Cuts tubing up to 1-1/2 in. (38 mm) diameter. Made of nickel-plated zinc-alloy die-casting.