Skip to Content
Merck
All Photos(1)

Key Documents

86893

Supelco

Tetrabutylammonium perchlorate

for electrochemical analysis, ≥99.0%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€76.10
50 G
€278.00

€76.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€76.10
50 G
€278.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(ClO4)
CAS Number:
Molecular Weight:
341.91
Beilstein:
3579274
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

€76.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0% (T)
≥99.0%

form

powder

mp

211-215 °C

solubility

acetonitrile: 0.1 g/mL, clear, colorless

SMILES string

[O-]Cl(=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.ClHO4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3,4)5/h5-16H2,1-4H3;(H,2,3,4,5)/q+1;/p-1

InChI key

KBLZDCFTQSIIOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium perchlorate (TBAP) is a quaternary ammonium salt, mainly used as electrolyte.
Visit our Sensor Applications portal to learn more.

Application

TBAP has been used as solvent in preparing standard stock solutions, for HPLC analysis of Sudan azo-dyes food samples.[1] It may be used as an electrophoretic buffer to rinse the capillary for the nonaqueous capillary electrophoresis (NACE) determination of atropine, anisodamine, and scopolamine in Flos daturae plant extract.[2]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of atropine, anisodamine, and scopolamine in plant extract by nonaqueous capillary electrophoresis coupled with electrochemiluminescence and electrochemistry dual detection.
Yuan B
Journal of Chromatography A, 1217(1), 171-174 (2010)
Analysis of sudan I, sudan II, sudan III, and sudan IV in food by HPLC with electrochemical detection: Comparison of glassy carbon electrode with carbon nanotube-ionic liquid gel modified electrode.
Chailapakul O
Food Chemistry, 109(4), 876-882 (2008)
David Balcells et al.
Dalton transactions (Cambridge, England : 2003), (30)(30), 5989-6000 (2009-07-23)
Experimental studies have shown that the C-H oxidation of Ibuprofen and methylcyclohexane acetic acid can be carried out with high selectivities using [(terpy')Mn(OH(2))(mu-O)(2)Mn(OH(2))(terpy')](3+) as catalyst, where terpy' is a terpyridine ligand functionalized with a phenylene linker and a Kemp's triacid
Benjamin T Diroll et al.
Nature communications, 10(1), 4511-4511 (2019-10-06)
Colloidal quantum wells are two-dimensional materials grown with atomically-precise thickness that dictates their electronic structure. Although intersubband absorption in epitaxial quantum wells is well-known, analogous observations in non-epitaxial two-dimensional materials are sparse. Here we show that CdSe nanoplatelet quantum wells
Atanu Jana et al.
Dalton transactions (Cambridge, England : 2003), 44(1), 359-367 (2014-11-11)
A novel electron rich, tetrathiafulvalene fused zinc porphyrin, (TTF)4PZn, has been newly synthesized and characterized using spectral and electrochemical methods. In spite of the presence of eight t-butyl groups, (TTF)4PZn exhibited appreciable aggregation in solution. Scanning electron microscopic (SEM) imaging

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service