Skip to Content
Merck
All Photos(1)

Key Documents

250228

Sigma-Aldrich

Tetrabutylammonium perrhenate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€107.00

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€107.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(ReO4)
CAS Number:
Molecular Weight:
492.67
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

Assay

98%

form

powder

SMILES string

[O-][Re](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.4O.Re/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;;;;/h5-16H2,1-4H3;;;;;/q+1;;;;-1;

InChI key

MTTXKKTWRLXAKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Starting material for rhenium complexes of o-phenylenediamine and o-aminobenzenethiol[1] and for perrhenate salt formation with nickel porphyrins.[2] Model compound for the synthesis of diagnostic radiopharmaceuticals.[3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Soc., Dalton Trans., 315-315 (1990)
Luo, L. et al.
Inorganic Chemistry, 32, 4491-4491 (1993)
Luo, H. et al.
Inorganic Chemistry, 34, 2287-2287 (1995)
Inorganic Chemistry, 29, 223-223 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service