Skip to Content
Merck
All Photos(1)

Documents

85795

Supelco

Vial

Selectophore

Synonym(s):

Bottle with conical shoulder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

product line

Selectophore

General description

5 mL, with ground glass stopper NS 12; borosilicate glass
Visit our Sensor Applications portal to learn more.

Application

for the dissolution of ion-selective membrane components during preparation of membranes for use in combination with glass plate (Cat. No. 48952) and glass rings (Cat. No. 48953, 48954)

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service