Skip to Content
Merck
All Photos(1)

Documents

45137

Supelco

Electrode Body ISE

Selectophore

Synonym(s):

Philips IS-561

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

product line

Selectophore

General description

Visit our Sensor Applications portal to learn more.

Application

Electrode Body ISE (Philips IS-561) was used to incorporate solvent polymeric membranes, during an experimental investigation done to study the correlation between the chemical structure and potentiometric ion-selectivity of biscrown ether derivatives.
For mounting of ion-selective membranes, electrode connecting wires (Cat. No. 45135, 45136) not included
Glass electrode spare part 74396

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Róbert Bereczki et al.
The Analyst, 130(1), 63-70 (2004-12-23)
The synthesis of a novel covalently immobilized crown ether based potassium ionophore is presented. Apart from previously proposed methods for the preparation of PVC linked ionophores based on the chemical modification of functionalized PVC polymers, the hereby proposed procedure involves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service