Skip to Content
Merck
All Photos(1)

Key Documents

85100

Sigma-Aldrich

Petrol

purum, bp 80-110 °C

Synonym(s):

Gasoline, Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
15101506
bp:
80-110 °C
Pricing and availability is not currently available.

grade

purum

Quality Level

form

liquid

bp

80-110 °C

density

0.70 g/mL at 20 °C

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Improved Gas Chromatographic Method for the Determination of C6-C12 Petroleum Hydrocarbons in Soil Water.
Voyevoda M, et al.
Clean: Soil, Air, Water, 36(2, 164-170 (2008)
GC methods for quantitative determination of benzene in gasoline.
Pavlova A and Ivanova R.
Acta Chromatographica , 215-225 (2003)
Hosein Karim et al.
Journal of injury & violence research, 7(2), 59-63 (2015-06-18)
To investigate factors related to prevention of self-immolation in west of Iran. In a case-control study, 30 consecutive cases of deliberate self-inflicted burns admitted to the regional burn centre (Imam Khomeini hospital in Kermanshah province, Iran) were compared with controls
J Järnberg et al.
The Science of the total environment, 199(1-2), 65-71 (1997-06-20)
The objective of this study was to compare the toxicokinetics of inhaled 1,2,4-trimethylbenzene (1,2,4-TMB) in man after exposure to white spirit with that observed after exposure to 1,2,4-TMB alone. TMBs occur mainly in petroleum products and the TMBs or their
I P Dick et al.
Human & experimental toxicology, 16(11), 645-651 (1998-01-14)
1. The absorption and distribution of lindane through skin was examined using human volunteers. Two different preparations were employed, one with acetone as the vehicle and the other, a commercial product, consisting primarily of white spirit as the solvent base.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service