Skip to Content
Merck
All Photos(1)

Key Documents

43728

Supelco

BTEX Solution

analytical standard

Synonym(s):

Benzene, Ethylbenzene, toluene, p-xylene, m-xylene, o-xylene Mix

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€68.40
10 X 1 ML
€191.00

€68.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€68.40
10 X 1 ML
€191.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€68.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.480

density

1.099 g/mL at 20 °C

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

BTEX are important chemicals, which are commonly used as chemical intermediates and solvents in the production of plastics.{11)
BTEX belongs to the family of organopollutants, which tend to accumulate in groundwater and contaminate the soil and air as well. These compounds are extremely toxic even in small concentrations and have been reported to cause permanent damage to the central nervous system.[1]

Application

BTEX solution has been used as a standard in the determination of BTEX in soil, using gas chromatography (GC) coupled with flame ionization detector.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

solution contains benzene, toluene, ethylbenzene, p-xylene, m-xylene, o-xylene, 1 mg/ml each in DMSO

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Field analysis of benzene, toluene, ethylbenzene and xylene in water by portable gas chromatography?microflame ionization detector combined with headspace solid-phase microextraction
Ji J, et al.
Talanta, 69, 894-899 (2006)
Determination of benzene, toluene, ethylbenzene and xylenes in soils by multiple headspace solid-phase microextraction
Ezquerro O, et al.
Journal of Chromatography A, 1035, 17-22 (2004)
Carla B Vidal et al.
Journal of environmental management, 112, 178-185 (2012-08-28)
Various technologies have been used for the treatment and remediation of areas contaminated by BTEX (benzene, toluene, ethylbenzene and xylenes), which are organic compounds that are of particular concern due to their toxicity. Potential applications of synthetic zeolites for environmental

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service