Skip to Content
Merck
All Photos(1)

Key Documents

77180

Millipore

Animal Proteins Peptone for Biotechnology and Microbiology

Enzymatically Digested, for Biotechnology and Microbiology

Synonym(s):

Peptone porcine

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€130.00

€130.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€130.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

€130.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Peptone from animal proteins, suitable for microbiology

biological source

Porcine skeletal muscle

Quality Level

form

powder

composition

total nitrogen (N), ≥10%

packaging

pkg of 100 g
pkg of 500 g

ign. residue

≤22%

loss

≤6% loss on drying

pH

7.0±0.5 (25 °C in H2O)

solubility

H2O: 2%, clear, faintly to light brownish-yellow

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Peptone from animal proteins is an enzymatic hydrolysate, obtained by digestion of lean meat with proteolytic enzymes, it is used as a nitrogen source in culture media formulated for isolating and cultivating fastidious and non-fastidious bacteria and fungi. In addition to its nitrogen constituents, this peptone has a high carbohydrate content and is suitable for many purposes. However, due to this high content of sugar, it is not recommendable for fermentation assays.

Application

Peptone is an enzymatic hydrolysate, obtained by digestion of lean meat with proteolytic enzymes is widely used in culture media and is often used for the cultivation of many fastidious organisms and where rapid, luxuriant growth is required.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Rojas et al.
Cell, 176(1-2), 306-317 (2018-12-07)
Trypanosome parasites control their virulence and spread by using quorum sensing (QS) to generate transmissible "stumpy forms" in their host bloodstream. However, the QS signal "stumpy induction factor" (SIF) and its reception mechanism are unknown. Although trypanosomes lack G protein-coupled
Federico Rojas et al.
Cell, 176(1-2), 306-317 (2018-12-07)
Trypanosome parasites control their virulence and spread by using quorum sensing (QS) to generate transmissible "stumpy forms" in their host bloodstream. However, the QS signal "stumpy induction factor" (SIF) and its reception mechanism are unknown. Although trypanosomes lack G protein-coupled

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service