66005BC
SimPlate® Total Coliform & E. coli
BioControl, Quantitative results for Coliforms and E. coli from food and environmental samples in just 24 hours
Synonym(s):
CEC
About This Item
Recommended Products
material
plastic vial (containing dehydrated culture media)
size
100 tests
form
dry powder
shelf life
6‑18 mo.
feature
fluorescent indicator
manufacturer/tradename
BioControl Systems 66005-100
BioControl Systems 66005-500
storage condition
dry media
application(s)
food and beverages
compatibility
for use with SimPlate (65009BC)
storage temp.
room temp
suitability
coliforms: Escherichia coli
General description
Features and Benefits
- Total Coliform and E. coli results are available from a single plate. Proprietary chromogenic media and patented plating device with isolation wells combine to produce easily counted results. Because results are not dependent on gas formation, results are more easily interpreted than tube or film-based methods.
- While other plating methods require duplicate plating of samples, and the standard MPN method requires multiple dilutions and at least nine tubes of media, SimPlate has been validated to provide equivalent results with just a single plate. Combined with the reduction of necessary sample dilutions, made possible by the broad counting range of the plate (1-738), SimPlate reduces the volume of media and number of tests required for each sample, helping to reduce overall testing costs.
Components
Testing Method
Other Notes
Legal Information
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service