Skip to Content
Merck
All Photos(1)

Key Documents

73043BC

Millipore

GDS Sample Wells Base

BioControl, For use with GDS

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€18.00

€18.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EACH
€18.00

About This Item

UNSPSC Code:
41104017

€18.00


Please contact Customer Service for Availability

Request more information

manufacturer/tradename

BioControl Systems 73043

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

Components

Bag of 10

Compatibility

BioControl GDS

Testing Method

PCR, Immunomagnetic Separation (IMS)

also commonly purchased with this product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service