63145
Magnesium sulfate heptahydrate
tested according to Ph. Eur.
Synonym(s):
Magnesii sulfas, Epsom salts
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Agency
USP/NF
tested according to Ph. Eur.
vapor density
<0.01 (vs air)
vapor pressure
<0.1 mmHg ( 20 °C)
SMILES string
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
InChI
1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2
InChI key
WRUGWIBCXHJTDG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pediatrics, 132(6), e1634-e1641 (2013-11-28)
Vaso-occlusive episodes (VOEs) are the most common complication of sickle cell disease in children. Treatment with magnesium seems to improve cellular hydration and may result in reduced vaso-occlusion. This study aimed to determine if intravenous (IV) magnesium sulfate (MgSO4) reduces
Pediatric blood & cancer, 61(6), 1049-1054 (2014-01-21)
Multiple recent Sickle Cell Disease studies have been terminated due to poor enrollment. We developed methods to overcome past barriers and utilized these to study the efficacy and safety of intravenous magnesium for vaso-occlusive crisis (VOC). We describe the methods
BJOG : an international journal of obstetrics and gynaecology, 121(5), 595-603 (2014-01-07)
To evaluate a slower (compared with a standard) infusion rate of the loading dose of magnesium sulphate for preterm fetal neuroprotection as a strategy to reduce maternal adverse effects. Randomised controlled trial. South Australian maternity hospital. Fifty-one women at <30
Best practice & research. Clinical obstetrics & gynaecology, 27(6), 877-884 (2013-08-22)
Hypertensive disorders of pregnancy are one of the leading causes of peripartum morbidity and mortality globally. Hypertensive disease in pregnancy is associated with a spectrum of severity, ranging from mild pregnancy-induced hypertension to eclampsia. Although most cases of pre-eclampsia may
Artificial organs, 37(1), 100-102 (2013-01-12)
Congenital heart disease (CHD) is relatively common and there are treatment options for virtually every form of CHD. With good care, the majority of babies can survive to adulthood; however, CHD remains a significant cause of death in infancy and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service