Merck
Severe pre-eclampsia and hypertensive crises.
Best practice & research. Clinical obstetrics & gynaecology (2013-08-22)
N Arulkumaran, L Lightstone
ABSTRACT
Hypertensive disorders of pregnancy are one of the leading causes of peripartum morbidity and mortality globally. Hypertensive disease in pregnancy is associated with a spectrum of severity, ranging from mild pregnancy-induced hypertension to eclampsia. Although most cases of pre-eclampsia may be managed successfully, severe pre-eclampsia is a life-threatening multisystem disease associated with eclampsia, HELLP (haemolysis, elevated liver enzymes, low platelets) syndrome, acute kidney injury, pulmonary oedema, placental abruption and intrauterine foetal death. Management of severe pre-eclampsia includes identification of high-risk patients, optimisation of antenatal care, early intervention and the identification and early management of complications. In the first instance, oral anti-hypertensive agents, including labetalol, nifedipine and methyldopa, should be tried. If oral anti-hypertensive agents have failed to adequately control blood pressure, intravenous anti-hypertensives should be considered. Commonly used intravenous anti-hypertensives include labetalol, hydralazine and glyceryl trinitrate. In addition to anti-hypertensive agents, close attention should be given to regular clinical examination, assessment of fluid balance, neurologic status and monitoring of other vital signs. Magnesium sulphate should be considered early to prevent seizures. Delivery of the baby is the definitive management of severe pre-eclampsia.
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Supelco
Trinitroglycerin solution, 1000 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Trinitroglycerin solution, 1 % (w/w) in propylene glycol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Hydralazine hydrochloride
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)