Skip to Content
Merck
All Photos(2)

Key Documents

63083

Supelco

Magnesium ionophore II

for ion-selective electrodes, Selectophore, function tested

Synonym(s):

ETH 5214, N,N′′-Octamethylene-bis(N′-heptyl-N′-methyl-methylmalonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H62N4O4
CAS Number:
Molecular Weight:
566.86
Beilstein:
6018631
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

quality

function tested

mp

63-65 °C

SMILES string

CCCCCCCN(C)C(=O)C(C)C(=O)NCCCCCCCCNC(=O)C(C)C(=O)N(C)CCCCCCC

InChI

1S/C32H62N4O4/c1-7-9-11-17-21-25-35(5)31(39)27(3)29(37)33-23-19-15-13-14-16-20-24-34-30(38)28(4)32(40)36(6)26-22-18-12-10-8-2/h27-28H,7-26H2,1-6H3,(H,33,37)(H,34,38)

InChI key

UTFJQHTUZRJEKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Other Notes

New synthetic neutral ionophore for magnesium. Assay of Mg2+-activity in intracellular liquids with microelectrodes based on Magnesium Ionophore II

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Buri et al.
Experimental physiology, 75(6), 751-761 (1990-11-01)
Intracellular free magnesium ([Mg2+]i) was measured in isolated ferret papillary muscles using ion-selective microelectrodes filled with the new magnesium sensor ETH 5214. This new sensor, unlike its predecessor ETH 1117, does not react to marked changes in K+, Na+ or
L A Blatter
Pflugers Archiv : European journal of physiology, 416(3), 238-246 (1990-05-01)
The application of a new type of intracellular magnesium-selective microelectrode based on the neutral carrier ETH5214 to measure intracellular free magnesium ([Mg]i) in frog skeletal muscle fibers is reported. At room temperature (18-20 degrees C) the average values for [Mg]i
Z M Hu et al.
Analytical chemistry, 61(6), 574-576 (1989-03-15)
A magnesium ion selective microelectrode based on a synthetic neutral carrier is presented. The selectivity of Mg2+ over Na+, K+, H+, and Ca2+ is sufficient for assays of intracellular magnesium ion activities. The microelectrodes with an optimized membrane composition have
Mathieu Métivier et al.
Current biology : CB, 31(4), 684-695 (2020-12-02)
Proper assembly of mitotic spindles requires microtubule nucleation not only at the centrosomes but also around chromatin. In this study, we found that the Drosophila tubulin-specific chaperone dTBCE is required for the enrichment of tubulin in the nuclear space after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service