Skip to Content
Merck
All Photos(1)

Key Documents

99409

Supelco

Magnesium ionophore II - cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
26111700
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

composition

Magnesium ionophore II, 10.0 wt. % (63083)
2-Nitrophenyl octyl ether, 87.0 wt. % (73732)
Potassium tetrakis(4-chlorophenyl)borate, 3.0 wt. %

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Other Notes

Assay of Mg2+ activity in intracellular liquids with microelectrodes†

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Z M Hu et al.
Analytical chemistry, 61(6), 574-576 (1989-03-15)
A magnesium ion selective microelectrode based on a synthetic neutral carrier is presented. The selectivity of Mg2+ over Na+, K+, H+, and Ca2+ is sufficient for assays of intracellular magnesium ion activities. The microelectrodes with an optimized membrane composition have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service