Skip to Content
Merck
All Photos(4)

Documents

55385

Supelco

3-Phosphoshikimic acid lithium salt

≥95.0% (HPLC)

Synonym(s):

Lithium 3-phosphoshikimate, Lithium shikimate-3-phosphate, S-3-P, S3P, Shikimic acid-3-phosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11O8P · xLi+
CAS Number:
Molecular Weight:
254.13 (free acid basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥95.0% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

OC(C1=C[C@@H](OP(O)(O)=O)[C@@H](O)[C@H](O)C1)=O

InChI

1S/C7H11O8P/c8-4-1-3(7(10)11)2-5(6(4)9)15-16(12,13)14/h2,4-6,8-9H,1H2,(H,10,11)(H2,12,13,14)/t4-,5-,6+/m1/s1

InChI key

QYOJSKGCWNAKGW-PBXRRBTRSA-N

Biochem/physiol Actions

Substrate of the pivotal EPSP synthase in the shikimate - chorismic acid pathway of most plants and many microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

From glucose to aromatics: recent developments in natural products of the shikimic acid pathway.
Ganem, B.
Tetrahedron, 32, 3353-3383 (1978)
Vanisree Mulabagal et al.
Analytical chemistry, 82(9), 3616-3621 (2010-04-17)
Shikimate kinase (SK) and other enzymes in the shikimate pathway are potential targets in the discovery of antimicrobial agents. In the current study, an ultrafiltration-liquid chromatography/mass spectrometry (UF-LC/MS) ligand based binding assay and an LC/MS based functional assay for Mycobacterium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service