Skip to Content
Merck
All Photos(1)

Documents

10108294001

Roche

Phosphoenol-pyruvate

97% (PEP-K), pkg of 1 g

Synonym(s):

Phospho(enol)pyruvic acid monopotassium salt, 2-(Phosphonooxy)-2-propenoic acid monopotassium salt, mono-Potassium phosphoenolpyruvate, PEP-K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4KO6P
CAS Number:
Molecular Weight:
206.13
Beilstein:
4603446
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

description

C3H4O6PK (Formula)
PEP, monopotassium salt

Quality Level

Assay

97% (PEP-K)

form

solid

mol wt

Mr 168.0 (PEP)
Mr 206.1 (PEP-K)

packaging

pkg of 1 g

manufacturer/tradename

Roche

storage temp.

2-8°C

SMILES string

[K+].OC(=O)C(=C)OP(O)([O-])=O

InChI

1S/C3H5O6P.K/c1-2(3(4)5)9-10(6,7)8;/h1H2,(H,4,5)(H2,6,7,8);/q;+1/p-1

InChI key

SOSDSEAIODNVPX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Contents: 80% PEP (enzymatic)

Application

Phosphoenolpyruvate (PEP) has been used in:
  • glutamate-cysteine ligase activity
  • v-ATPase enzyme activity assay
  • meso kinase assay

Substrate for pyruvate kinase.

Biochem/physiol Actions

Phosphoenolpyruvate (PEP) plays a vital role in metabolism. It has a high phosphate group-transfer potential and it serves a versatile C3-synthon in C–C, C–P and C–O bond-formation reactions. PEP acts as a substrate for pyruvate kinase (PK) in the last step of glycolysis.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of phosphoenol pyruvate (PEP) analogues and evaluation as inhibitors of PEP-utilizing enzymes.
Garcia A, et al.
FEBS Journal, 269(13), 3226-3236 (2002)
Glutathione participates in the modulation of starvation-induced autophagy in carcinoma cells.
Desideri E
Autophagy, 8(12), 1769-1781 (2012)
Jasmine M Hershewe et al.
Nature communications, 12(1), 2363-2363 (2021-04-24)
Cell-free gene expression (CFE) systems from crude cellular extracts have attracted much attention for biomanufacturing and synthetic biology. However, activating membrane-dependent functionality of cell-derived vesicles in bacterial CFE systems has been limited. Here, we address this limitation by characterizing native
Maria B Bagh et al.
Nature communications, 8, 14612-14612 (2017-03-08)
Defective lysosomal acidification contributes to virtually all lysosomal storage disorders (LSDs) and to common neurodegenerative diseases like Alzheimer's and Parkinson's. Despite its fundamental importance, the mechanism(s) underlying this defect remains unclear. The v-ATPase, a multisubunit protein complex composed of cytosolic
Brian S Muntean et al.
Cell reports, 34(5), 108718-108718 (2021-02-04)
The G protein alpha subunit o (Gαo) is one of the most abundant proteins in the nervous system, and pathogenic mutations in its gene (GNAO1) cause movement disorder. However, the function of Gαo is ill defined mechanistically. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service