Skip to Content
Merck
All Photos(1)

Key Documents

50195

Millipore

Listeria UVM I Supplement

suitable for microbiology

Synonym(s):

Antibiotic supplement, Listeria primary selective enrichment supplement (UVM I)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 5 vials

application(s)

environmental
food and beverages
pathogen testing

microbiology

storage temp.

2-8°C

suitability

Listeria spp.

General description

Listeria UVM I Supplement is used as an antibiotic supplement in culture media to inhibit the growth of unwanted organisms while allowing the growth of the desired microorganisms. This antibiotic supplement is recommended for the selective isolation and enumeration of Listeria sp. The supplement consists of a combination of Acriflavin hydrochloride (6 mg) and Nalidixic (10 mg) with broad-spectrum antimicrobial activity against various Gram-positive and Gram-negative organisms thereby selectively allowing the growth of Listeria sp.

Application

Listeria UVM I Supplement is an antibiotic supplement that is recommended for the selective isolation and enumeration of Listeria species from various food and clinical samples.

Components

Composition:
(per vial, sufficient for 500 ml medium)
Acriflavin hydrochloride 6.0 mg
Nalidixic 10.0 mg

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service