Skip to Content
Merck
All Photos(1)

Key Documents

46525

Supelco

19-Norethindrone

VETRANAL®, analytical standard

Synonym(s):

17α-Ethynyl-19-nortestosterone, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one, 19-Nor-17α-ethynyl-4-androsten-17β-ol-3-one, 19-Norethisterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26O2
CAS Number:
Molecular Weight:
298.42
Beilstein:
1915671
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

205-206 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12CCC(=O)C=C1CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@]4(O)C#C

InChI

1S/C20H26O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,12,15-18,22H,4-11H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1

InChI key

VIKNJXKGJWUCNN-XGXHKTLJSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

19-norethindrone is an oral contraceptive involved in the inhibition of cytosolic sulfotransferases (SULT).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karl van den Broeck et al.
The European journal of contraception & reproductive health care : the official journal of the European Society of Contraception, 17(5), 321-328 (2012-09-15)
Ferdinand Peeters, a practising gynaecologist from Turnhout, near Antwerp (Belgium), had a pivotal role in the development of a practical and viable modality of oral contraception, striking the balance between efficacy and an acceptably low incidence and severity of side
Isabelle Demeestere et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(7), 903-909 (2012-11-07)
To assess the efficacy of gonadotropin-releasing hormone agonist (GnRHa) in preventing chemotherapy-induced ovarian failure in patients treated for Hodgkin or non-Hodgkin lymphoma within the setting of a multicenter, randomized, prospective trial. Patients age 18 to 45 years were randomly assigned
V Koskela-Niska et al.
Climacteric : the journal of the International Menopause Society, 16(1), 48-53 (2012-05-30)
While previous data link the use of postmenopausal hormone therapy to an increased risk for ovarian cancer, little is known about the impact of various progestins, modes or routes of administration of hormone therapy for this risk. In this nationwide
Louise Lind Schierbeck et al.
BMJ (Clinical research ed.), 345, e6409-e6409 (2012-10-11)
To investigate the long term effect of hormone replacement therapy on cardiovascular outcomes in recently postmenopausal women. Open label, randomised controlled trial. Denmark, 1990-93. 1006 healthy women aged 45-58 who were recently postmenopausal or had perimenopausal symptoms in combination with
Patricia M LoRusso et al.
Cancer chemotherapy and pharmacology, 71(1), 193-202 (2012-10-16)
Vismodegib, a first-in-class oral hedgehog pathway inhibitor, is an effective treatment for advanced basal cell carcinoma. Based on in vitro data, a clinical drug-drug interaction (DDI) assessment of cytochrome P450 (CYP) 2C8 was necessary; vismodegib's teratogenic potential warranted a DDI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service