Skip to Content
Merck
All Photos(5)

Key Documents

1.07017

Supelco

Ethyl alcohol, pure

greener alternative

≥99.5% (GC), EMPARTA® ACS reagent, for extraction and analytical purposes

Synonym(s):

Ethanol, Ethyl alcohol, EtOH, Ethyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€75.60
4 L
€133.00
25 L
€450.00

€75.60


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
2.5 L
€75.60
4 L
€133.00
25 L
€450.00

About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-578-6
NACRES:
NB.21
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)
59 hPa ( 20 °C)

€75.60


Estimated to ship on07 April 2025Details


Request a Bulk Order

Product Name

Ethanol absolute, for analysis EMPARTA® ACS

grade

ACS reagent
reagent grade
absolute

Quality Level

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)
59 hPa ( 20 °C)

description

for analytical purposes
for extraction

product line

EMPARTA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

683 °F

potency

6200 mg/kg LD50, oral (Rat)

expl. lim.

19 %, 60 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

dilution

(for routine analytical applications)

impurities

≤0.0002% Substances reducing potassium permanganate (as O)
≤0.0002 meq/g Titrable base
≤0.0005 meq/g Titrable acid
≤0.001% Acetone (GC)
≤0.01% 2-Propanol (GC)
≤0.1% Methanol (GC)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−114 °C (lit.)

transition temp

flash point 12 °C

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

greener alternative category

storage temp.

2-30°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMD Millipore offers EMPARTA grade solvents for routine tasks in analytical laboratories. EMPARTA grade solvents offer fewer test parameters than EMSURE, yet are still of high quality. EMPARTA solvents meet ACS requirements, making them ideal for a wide range of analytical applications.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biorenewable and alternative to petrochemical solvents. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here  for more information.

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Color: ≤ 10 Hazen
Solubility in water: conforms
Titrable acid: ≤ 0.0005 meq/g
Titrable base: ≤ 0.0002 meq/g
Acetone (GC): ≤ 0.001 %
Methanol (GC): ≤ 0.1 %
2-Propanol (GC): ≤ 0.01 %
Acetone, Isopropyl Alcohol (ACS): conforms
Substances reducing potassium permanganate (as O): ≤ 0.0002 %
Substances reducing permanganate (ACS): conforms
Readily carbonizable substances: conforms
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.2 %

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrique Ros et al.
Lab on a chip, 20(5), 958-972 (2020-01-29)
Cell migration is a key function in a myriad of physiological events and disease conditions. Efficient, quick and descriptive profiling of migration behaviour in response to different treatments or conditions is highly desirable in a series of applications, ranging from

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service