Skip to Content
Merck
All Photos(1)

Key Documents

46092

Supelco

[12-(4-Ethylphenyl)dodecyl] 2-nitrophenyl ether

Selectophore, ≥95.0%

Synonym(s):

ETH 8045

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37NO3
CAS Number:
Molecular Weight:
411.58
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥95.0%

density

1.022 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCc1ccc(CCCCCCCCCCCCOc2ccccc2[N+]([O-])=O)cc1

InChI

1S/C26H37NO3/c1-2-23-18-20-24(21-19-23)15-11-9-7-5-3-4-6-8-10-14-22-30-26-17-13-12-16-25(26)27(28)29/h12-13,16-21H,2-11,14-15,22H2,1H3

InChI key

KTBRGUBCCOJFFI-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.
[12-(4-Ethylphenyl)dodecyl] 2-nitrophenyl ether is a plasticizer mainly used during electrode preparation.[1]

Application

It was used in preparation of PVC membrane cocktail with multi-wall carbon nanotubes, in a study done to investigate the use of solid-contact (SC) ion-selective electrodes (ISE) in combination with an radio-frequency identification potentiometer.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lipophilic plasticizer (log PTLC=12.8) with excellent characteristics for use in magnesium selective membrane electrodes[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Novell et al.
The Analyst, 138(18), 5250-5257 (2013-07-17)
Instrumental approaches to remotely and wirelessly monitoring chemical species are increasingly needed. Together with the electronic developments, efforts to optimize and validate the performance of these new devices are required. In this work, the analytical performance of a recently developed
A Comparison of Neutral Mg2+-Selective Ionophores in Solvents Polymeric Membranes: Complex Stoichiometry
Zhang, W., et al.
Analytical Sciences, 16, 11-18 (2000)
R. Eugster et al.
Analytica Chimica Acta, 289, 1-1 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service