Skip to Content
Merck
All Photos(1)

Key Documents

44224

Supelco

Dodecyl 2-nitrophenyl ether

Selectophore, ≥99.0%

Synonym(s):

1-Dodecyloxy-2-nitrobenzene, 2-Nitrophenyl dodecyl ether, ETH 217

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€182.00

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€182.00

About This Item

Empirical Formula (Hill Notation):
C18H29NO3
CAS Number:
Molecular Weight:
307.43
Beilstein:
3350321
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

product line

Selectophore

Quality Level

Assay

≥99.0% (GC)
≥99.0%

refractive index

n20/D 1.503

bp

218 °C/1.5 mmHg (lit.)

density

0.999 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCOc1ccccc1[N+]([O-])=O

InChI

1S/C18H29NO3/c1-2-3-4-5-6-7-8-9-10-13-16-22-18-15-12-11-14-17(18)19(20)21/h11-12,14-15H,2-10,13,16H2,1H3

InChI key

DXCTUMLYTQODSP-UHFFFAOYSA-N

General description

Dodecyl 2-nitrophenyl ether is an nitro-aromatic solvent.[1]
Visit our Sensor Applications portal to learn more.

Application

It was used to enhance extraction of nonpolar drugs during a study to demonstrate different approaches of basic drugs to maximize extraction recovery using electro membrane extraction method.[1]

Other Notes

Plasticizer for solvent polymeric membranes in ion-selective electrodes[2][3]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuto Osaki et al.
Sensors (Basel, Switzerland), 19(12) (2019-06-20)
We have investigated human-stress monitoring by making use of salivary nitrate, which can be a candidate for stress markers, with ion-selective field-effect transistors (ISFETs). ISFETs are suitable for on-site single-drop analysis of salivary nitrate within 10 s. However, when ISFETs
R. Yuan et al.
Analytical Chemistry, 65, 2572-2572 (1993)
Torunn M Middelthon-Bruer et al.
Journal of separation science, 31(4), 753-759 (2008-02-02)
Thirty-five different basic drugs were extracted by electro membrane extraction (EME), from acidified samples containing HCl as the BGE, through an organic solvent immobilized in the pores in the wall of a porous hollow fiber (supported liquid membrane, SLM), and
K. Fluri et al.
Helvetica Chimica Acta, 75, 1012-1012 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service