Skip to Content
Merck
All Photos(1)

Documents

44542

Supelco

γ-Nonalactone

analytical standard

Synonym(s):

γ-Nonanoic lactone, Coconut aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.446-1.450
n20/D 1.447 (lit.)

bp

121-122 °C/6 mmHg (lit.)

density

0.976 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCC1CCC(=O)O1

InChI

1S/C9H16O2/c1-2-3-4-5-8-6-7-9(10)11-8/h8H,2-7H2,1H3

InChI key

OALYTRUKMRCXNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Nonalactone is a flavor volatile compound, belonging to the group of lactones and is one of the minor chiral compound present in vanilla.

Application

γ-Nonalactone may be used as an analytical reference standard for the identification and quantification of the analyte in freshly cooked non-scented rice using modified headspace solid-phase microextraction (SPME) method followed by gas chromatography-olfactometry (GC-O) analysis and gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Screening for gamma-nonalactone in the headspace of freshly cooked non-scented rice using SPME/GC-O and SPME/GC-MS
Zheng Z, et al.
Molecules (Basel), 14(8), 2927-2934 (2009)
Medicinal and Aromatic Plants - Industrial Profiles
Vanilla (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service