Skip to Content
Merck
All Photos(4)

Key Documents

W278140

Sigma-Aldrich

γ-Nonanoic lactone

greener alternative

natural, 98%, FG

Synonym(s):

Aldehyde C18, Coconut aldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

W278140-SAMPLE-K
€49.30
100 G
€115.00
1 KG
€441.00
5 KG
€1,810.00

€49.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W278140-SAMPLE-K
€49.30
100 G
€115.00
1 KG
€441.00
5 KG
€1,810.00

About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
FEMA Number:
2781
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.001
NACRES:
NA.21
Organoleptic:
coconut; creamy; fatty; oily; buttery; sweet
grade:
FG
Fragrance grade
Halal
Kosher
natural
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

€49.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.447 (lit.)

bp

121-122 °C/6 mmHg (lit.)

density

0.976 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

coconut; creamy; fatty; oily; buttery; sweet

SMILES string

CCCCCC1CCC(=O)O1

InChI

1S/C9H16O2/c1-2-3-4-5-8-6-7-9(10)11-8/h8H,2-7H2,1H3

InChI key

OALYTRUKMRCXNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application

In flavor applications, this compound contributes to caramel notes. In fragrance applications, this compound contributes is used in amber, balsam, floral, juniper berry, nutmeg, pine, coconut, maple and oriental notes.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raw Material Bulletin
Perfumer & Flavorist, 39(3), 18-18 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service