Skip to Content
Merck
All Photos(1)

Documents

43080

Sigma-Aldrich

1,1-Diphenylhydrazine hydrochloride

technical, ≥97.0% (AT)

Synonym(s):

N,N-Diphenylhydrazine (asym.) hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NNH2 · HCl
CAS Number:
Molecular Weight:
220.70
Beilstein:
3569340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥97.0% (AT)

mp

162-166 °C (dec.)

SMILES string

Cl.NN(c1ccccc1)c2ccccc2

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service