193933
Hexacyclen trisulfate
Synonym(s):
1,4,7,10,13,16-Hexaazacyclooctadecane trisulfate
Select a Size
About This Item
Recommended Products
form
solid
SMILES string
OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.C1CNCCNCCNCCNCCNCCN1
InChI
1S/C12H30N6.3H2O4S/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1;3*1-5(2,3)4/h13-18H,1-12H2;3*(H2,1,2,3,4)
InChI key
UXVDKNLOZWKLJC-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service