Skip to Content
Merck
All Photos(2)

Documents

42206

Supelco

Flumetsulam

PESTANAL®, analytical standard

Synonym(s):

2′,6′-Difluoro-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonanilide, N-(2,6-Difluorophenyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9F2N5O2S
CAS Number:
Molecular Weight:
325.29
Beilstein:
8155688
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 43.4-45.2 wt. %
nitrogen content, 21.1-22.0 wt. %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1ccn2nc(nc2n1)S(=O)(=O)Nc3c(F)cccc3F

InChI

1S/C12H9F2N5O2S/c1-7-5-6-19-11(15-7)16-12(17-19)22(20,21)18-10-8(13)3-2-4-9(10)14/h2-6,18H,1H3

InChI key

RXCPQSJAVKGONC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flumetsulam is postemergence, triazolopyrimidine sulfonanilide herbicide, which can find applications in agriculture, for under-sown wheat and certain legume crops and pastures. Its mode of action involves the suppression of acetolactate synthase, thereby inhibiting the amino acid synthesis in plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption isotherm modeling of carbendazim and flumetsulam onto homoionic-montmorillonite clays: comparison of linear and nonlinear models
Maataoui EY, et al.
Turkish Journal of Chemistry (2017)
Alkoxy base-mediated selective synthesis and new rearrangements of 1, 2, 4-triazolodipyrimidinones
Pyatakov.AD, et al.
Tetrahedron Letters, 58, 748-754 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service