Skip to Content
Merck
All Photos(1)

Key Documents

32203-M

Sigma-Aldrich

Diethyl ether

Diethyl ether

≥99.8% (GC), ACS reagent, puriss. p.a., contains BHT as inhibitor, reag. Ph. Eur., reag. ISO, USP/NF

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€72.20
2.5 L
€158.00

€72.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€72.20
2.5 L
€158.00

About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
34.6 °C (lit.)

€72.20


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

2.6 (vs air)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

320 °F

contains

BHT as inhibitor

expl. lim.

36.5 %

technique(s)

GC/GC: suitable

impurities

≤0.00007% adehydes (as HCHO)
≤0.0002% free acid (as CH3COOH)
≤0.001% carbonyl compounds (as CO)
≤0.001% non-volatile matter
≤0.005% acetone (GC)
≤0.02% ethanol (GC)
≤0.3 ppm peroxides (as H2O2)
0.03% water (Karl Fischer)
5-8 ppm BHT (GC)

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.05 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.05 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.05 mg/kg

SMILES string

CCOCC

suitability

passes test for reaction against H2SO4

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl ether (DEE) is widely employed as solvent for Grignard synthesis.[1] Its efficacy as solvent for formalin-ether (F-E) sedimentation technique has been studied.[2] It has been reported as a hydroboration solvent.[3] DEE is a promising compression-ignition fuel and its energy density is much higher than ethanol. It is also one of the components in a blend used in diesel fuel. It can also be used as alternative of diesel fuel.[4]

Application

Diethyl ether may be employed as solvent for the oxidation of secondary alcohols into ketones.[3] It may be employed as solvent in the preparation of triblock copolymer of PCL-PEG-PCL, acrylated PCL-PEG-PCL and the pentablock copolymer PAE-PCL-PEG-PCL-PAE [PAE = poly(β-amino ester), PEG = poly(ethylene glycol), PCL = poly(ε-caprolactone)].[5]

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F

Flash Point(C)

-40.00 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dai P Huynh et al.
Biomaterials, 29(16), 2527-2534 (2008-03-11)
The concept of this research is using poly(beta-amino ester) (PAE) as a duo-functional group for synthesis of the novel sensitive injectable hydrogel for controlled drug/protein delivery. Firstly, PAE made of 1,4-butanediol diacrylate and 4,4'-trimethylene dipiperidine is used as a pH-sensitive
K H Young et al.
Journal of clinical microbiology, 10(6), 852-853 (1979-12-01)
Ethyl acetate appears to be a satisfactory subsitute solvent for diethyl ether in the Formalin-ether sedimentation technique. In comparative studies, concentration of organisms with ethyl acetate was equal to or greater than that with diethyl ether. No distortion or alteration

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service