Skip to Content
Merck
All Photos(2)

Key Documents

32427

Supelco

Orthosulfamuron

PESTANAL®, analytical standard

Synonym(s):

1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[2-(dimethylcarbamoyl)phenylsulfamoyl]urea

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€347.00

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€347.00

About This Item

Empirical Formula (Hill Notation):
C16H20N6O6S
CAS Number:
Molecular Weight:
424.43
Beilstein:
11343666
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(OC)nc(NC(=O)NS(=O)(=O)Nc2ccccc2C(=O)N(C)C)n1

InChI

1S/C16H20N6O6S/c1-22(2)14(23)10-7-5-6-8-11(10)20-29(25,26)21-16(24)19-15-17-12(27-3)9-13(18-15)28-4/h5-9,20H,1-4H3,(H2,17,18,19,21,24)

InChI key

UCDPMNSCCRBWIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Orthosulfamuron is a systemic post-emergence herbicide belonging to the sulfamoylurea class of herbicides. Sulfamoylurea herbicides act through inhibition of branch-chain amino acid biosynthesis in plants. Orthosulfamuron′s specific mode of action is through the inhibition of the plant enzyme, acetolactate synthase, also known as acetohydroxy acid synthase (ALS/AHAS). Inhibition of ALS/AHAS blocks the branched-chain amino acid biosynthesis of valine, leucine, and isoleucine, which are involved in the plant growth process. Orthosulfamuron can be synthesized by reaction of N,N-dimethyl 2-sulfamoylchloride benzcarbamate with 2-amino-4,6-dimethoxypyrimidine in the presence of a base.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rohit Pandey et al.
Environmental monitoring and assessment, 192(Suppl 1), 813-813 (2021-01-15)
Orthosulfamuron, a rice herbicide of the family pyrimidinyl sulfonylurea, causes ground water contamination for its moderate persistence and high water solubility. It may cause phytotoxicity and, thereby, affect the growth of succeeding crops. Keeping in view of the environmental safety

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service