Skip to Content
Merck
All Photos(1)

Key Documents

21065

Millipore

Calcium caseinate Agar

NutriSelect® Basic, suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€262.00

€262.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€262.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€262.00


Please contact Customer Service for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 13 g/L
calcium caseinate, 3.5 g/L
calcium chloride dihydrate, 0.2 g/L
casein peptone, 2.0 g/L
disodium hydrogen phosphate, 0.105 g/L
meat extract, 2.0 g/L
meat peptone, 4.0 g/L
potassium dihydrogen phosphate, 0.035 g/L
sodium chloride, 5.0 g/L
tripotassium citrate monohydrate, 0.35 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

nonselective and differential for Bacillus cereus
nonselective and differential for Bacillus spp.
nonselective and differential for Pseudomonas spp.
nonselective and differential for bacteria (General Media)
proteolytic bacteria

Application

Selective agar for the detection and enumeration of protein metabolising microorganisms (proteolytes) in food according to Frazier and Rupp (modified).

Preparation Note

Dissolve 30.7 g in 1 litre distilled water and autoclave at 121°C for 15 minutes.

Other Notes

For protease testing in an identification scheme for coagulase negative staphylococci from bovine mastitis[1]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Devriese et al.
Research in veterinary science, 57(2), 240-244 (1994-09-01)
Coagulase-negative staphylococci from cases of bovine mastitis were identified to species level by using an identification scheme based on a three-plate test system which tested for DNase on DNA agar, for protease on calcium caseinate agar, and for the organism's

Articles

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service