Skip to Content
Merck
All Photos(1)

Key Documents

18038

Millipore

Fraser Selective Supplement

suitable for microbiology

Synonym(s):

Listeria Selective Supplement acc. to Fraser

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
€56.20

€56.20


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 VIALS
€56.20

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

€56.20


Please contact Customer Service for Availability

Request more information

Agency

according to GB 4789.30-2016

Quality Level

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages
pathogen testing

microbiology

suitability

Listeria spp.

General description

Recommended by ISO Committee for Listeria monocytogens from food, animal feed etc.

Application

for Listeria monocytogens from food, animal feed etc.

Components

(per vial, sufficient for 500 ml medium)
Nalidixic acid 10.0 mg
Acriflavin Hydrochloride 12.5 mg

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service