Recommended Products
grade
ACS spectrophotometric grade
Quality Level
vapor density
2 (vs air)
vapor pressure
184 mmHg ( 20 °C)
Assay
≥99.5%
form
liquid
expl. lim.
13.2 %
technique(s)
UV/Vis spectroscopy: suitable
impurities
MnO4 reducers, passes test
≤0.0003 meq/g Titr. acid
≤0.0006 meq/g Titr. base
≤0.002% acetaldehyde
≤0.05% isopropanol
≤0.05% methanol
≤0.5% water
evapn. residue
<0.001%
refractive index
n20/D 1.359 (lit.)
bp
56 °C/760 mmHg (lit.)
mp
−94 °C (lit.)
density
0.791 g/mL at 25 °C (lit.)
λ
H2O reference
UV absorption
λ: 330 nm Amax: 1.00
λ: 340 nm Amax: 0.10
λ: 350 nm Amax: 0.02
λ: 400 nm Amax: 0.01
format
neat
SMILES string
CC(C)=O
InChI
1S/C3H6O/c1-3(2)4/h1-2H3
InChI key
CSCPPACGZOOCGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.0 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service