Skip to Content
Merck
All Photos(2)

Key Documents

1.00022

Supelco

Acetone

for spectroscopy Uvasol®

Synonym(s):

Dimethyl ketone, Propanone, 2-Propanone

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€67.90
2.5 L
€243.00

€67.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 ML
€67.90
2.5 L
€243.00

About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-662-2
NACRES:
NA.03
grade:
spectrophotometric grade
technique(s):
UV/Vis spectroscopy: suitable
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

€67.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

spectrophotometric grade

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

product line

Uvasol®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

465 °C (DIN 51794)

potency

5800 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

expl. lim.

13.2 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

330 nm, ≥15%
335 nm, ≥60%
340 nm, ≥85%
345 nm, ≥95%
350 nm, ≥99%

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

capacity

1000  g/kg water absorbing capacity

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

UV absorption

λ: 330 nm Amax: ≤0.82
λ: 335 nm Amax: ≤0.22
λ: 340 nm Amax: ≤0.07
λ: 345 nm Amax: ≤0.02
λ: 350 nm Amax: ≤0.004

format

neat

storage temp.

2-30°C

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service