Skip to Content
Merck
All Photos(1)

Key Documents

03629

Supelco

5-Ethyl-2,3-dimethylpyrazine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
Beilstein:
956766
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.498-1.502

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCC1=NC(C)=C(C)N=C1

InChI

1S/C8H12N2/c1-4-8-5-9-6(2)7(3)10-8/h5H,4H2,1-3H3

InChI key

CIBKSMZEVHTQLG-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Yan et al.
Foods (Basel, Switzerland), 10(2) (2021-03-07)
Pyrazines are important compounds in soy sauce aroma type Baijiu (SSAB). In this work, a total of 16 pyrazines were analyzed using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS) in SSAB. The quantitative results showed that 2,3,5,6-tetramethylpyrazine, 2,6-dimethylpyrazine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service