Skip to Content
Merck
All Photos(1)

Key Documents

94560

Supelco

Methyl valerate

analytical standard

Synonym(s):

Methyl pentanoate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€53.30
5 ML
€122.00

€53.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€53.30
5 ML
€122.00

About This Item

Linear Formula:
CH3(CH2)3CO2CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741905
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€53.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.8% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.397 (lit.)
n20/D 1.398

bp

128 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCC(=O)OC

InChI

1S/C6H12O2/c1-3-4-5-6(7)8-2/h3-5H2,1-2H3

InChI key

HNBDRPTVWVGKBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl valerate is a methyl ester, which may find application as a surrogate component in the production of biodiesel.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experiments and modeling of the autoignition of methyl pentanoate at low to intermediate temperatures and elevated pressures in a rapid compression machine
Weber.WB, et al.
Fuel: The Science and Technology of Fuel and Energy, 212, 479-486 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service