Skip to Content
Merck
All Photos(1)

Key Documents

24-6660

Sigma-Aldrich

Pyrogallol

JIS special grade, ≥99.0%

Synonym(s):

1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, Pyrogallic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
Beilstein:
907431
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
grade:
JIS special grade
Assay:
≥99.0%
technique(s):
toxicology assay: suitable
bp:
309 °C (lit.)
vapor pressure:
10 mmHg ( 167.7 °C)
Pricing and availability is not currently available.

grade

JIS special grade

vapor density

4.4 (vs air)

vapor pressure

10 mmHg ( 167.7 °C)

Assay

≥99.0%

form

crystalline

availability

available only in Japan

technique(s)

toxicology assay: suitable

pH

5.8 (10 g/L)

bp

309 °C (lit.)

mp

133-134 °C (lit.)

solubility

water: soluble

SMILES string

Oc1cccc(O)c1O

InChI

1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H

InChI key

WQGWDDDVZFFDIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeannette Geschwind et al.
Macromolecular rapid communications, 34(2), 150-155 (2012-11-30)
A series of functional polycarbonates, poly((isopropylidene glyceryl glycidyl ether)-co-(glycidyl methyl ether) carbonate) (P((IGG-co-GME) C)) random copolymers with different fractions of 1,2-isopropylidene glyceryl glycidyl ether (IGG) units, is synthesized. After acidic hydrolysis of the acetal protecting groups, a new type of
Adelaide U P Hain et al.
Journal of structural biology, 180(3), 551-562 (2012-09-18)
The autophagy-related proteins are thought to serve multiple functions in Plasmodium and are considered essential to parasite survival and development. We have studied two key interacting proteins, Atg8 and Atg3, of the autophagy pathway in Plasmodium falciparum. These proteins are
Drew A Fowler et al.
Chemical communications (Cambridge, England), 49(18), 1802-1804 (2013-01-30)
The crystal structure of a cocrystal comprising a complex of a bis(imidazolium) ionic liquid hosted by pyrogallol[4]arene (PgC) macrocycles is reported. Introduction of the dicationic species yields complexation by two PgC macrocycles, the resulting bilayer structure formed from alternately arranged
Jennifer C Chapin et al.
Journal of the American Chemical Society, 134(36), 15000-15009 (2012-09-04)
Pyrogallol[4]arene is a macrocycle with a concave surface and 12 peripheral hydroxyl groups that mediate its self-assembly to form hexamers of octahedral symmetry in the solid state, in solution, and in the gas phase. These hexamers enclose approximately 1300 Å(3)
M Buchweitz et al.
Food chemistry, 135(4), 3010-3019 (2012-09-18)
The formation of anthocyanin-metal chelates, exhibiting intense blue colours was monitored over a period up to 10 weeks. Evaluating normalised absorption spectra in the range of 580-700 nm and their proportion of the total area under the curve (AUC), provided

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service