Merck
  • Formation of a dimeric host-guest complex via binding between a dicationic ionic liquid and a pyrogallol[4]arene macrocycle.
Formation of a dimeric host-guest complex via binding between a dicationic ionic liquid and a pyrogallol[4]arene macrocycle.
Chemical communications (Cambridge, England) (2013-01-30)
Drew A Fowler, Jerry L Atwood, Gary A Baker
ABSTRACT
The crystal structure of a cocrystal comprising a complex of a bis(imidazolium) ionic liquid hosted by pyrogallol[4]arene (PgC) macrocycles is reported. Introduction of the dicationic species yields complexation by two PgC macrocycles, the resulting bilayer structure formed from alternately arranged dimeric host-guest complexes.
Product Description
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)