24-5720
Potassium thiocyanate
SAJ first grade, ≥99.0%
Synonym(s):
Potassium rhodanide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
SAJ first grade
Assay
≥99.0%
form
solid
availability
available only in Japan
pH
5.3-8.7 (25 °C, 97.2 g/L)
mp
173 °C (lit.)
SMILES string
[K]SC#N
InChI
1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1
InChI key
ZNNZYHKDIALBAK-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1
Supplementary Hazards
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The journal of physical chemistry. B, 113(29), 10014-10021 (2009-07-03)
The results of a neutron diffraction experiment performed on aqueous solutions of KSCN are analyzed and discussed in comparison with similar data for KCl and KOH solutions. The effect of the different ions on the structure of water and the
The Journal of organic chemistry, 77(8), 4148-4151 (2012-03-27)
A concise approach to various benzisothiazol-3(2H)-one derivatives has been developed by copper-catalyzed the reaction of o-bromobenzamide derivatives with potassium thiocyanate (KSCN) in water. The reaction proceeds via a tandem reaction with S-C bond and S-N bond formation.
Reproductive toxicology (Elmsford, N.Y.), 23(4), 568-577 (2007-03-06)
A study was performed at term and at weaning to verify the toxic effects of the prenatal exposure to potassium cyanide (KCN) and potassium thiocyanate (KSCN) in pregnant Wistar rats. Females received daily in drinking water the doses: 1, 3
The journal of physical chemistry. B, 116(49), 14426-14432 (2012-11-28)
Microscopic structures and dynamics of aqueous salt solutions were investigated with the ultrafast vibrational energy exchange method and anisotropy measurements. In KSCN aqueous solutions of various concentrations, the rotational time constants of SCN(-) anions are proportional to the viscosities of
Human vaccines, 2(4), 174-180 (2006-10-03)
The major surface antigen of Hepatitis B virus (HBsAg) is a cysteine-rich, lipid-bound protein with 226 amino acids. Recombinant HBsAg (rHBsAg) with associated lipids can self-assemble into 22-nm immunogenic spherical particles, which are used in licensed Hepatitis B vaccines. Little
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service