Skip to Content
Merck

1.06345

SAFC

Sodium dihydrogen phosphate dihydrate

EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JPE, USP, E 339

Pharma Manufacturing

Synonym(s):

Sodium phosphate monobasic dihydrate, mono-Sodium orthophosphate, Sodium biphosphate, Sodium phosphate monobasic, Sodium dihydrogen phosphate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€35.00
5 KG
€69.00
12 KG
€166.00
25 KG
€291.00
25 KG
€348.00

€35.00


Estimated to ship on07 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€35.00
5 KG
€69.00
12 KG
€166.00
25 KG
€291.00
25 KG
€348.00

About This Item

Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
MDL number:
UNSPSC Code:
12352302
E Number:
E339
EC Index Number:
231-449-2

€35.00


Estimated to ship on07 April 2025Details


Request a Bulk OrderRequest a Sample

Agency

BP
ChP
JPE
Ph. Eur.
USP

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

pH

4.5 (25 °C, 12 g/L in H2O)

mp

60 °C

solubility

850 g/L

density

1.915 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-30°C

SMILES string

O.O.[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+1;;;/p-1

InChI key

VBJGJHBYWREJQD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium dihydrogen phosphate dihydrate is typically used as a phosphate buffer system component for biomolecule downstream chromatography steps or final liquid formulation.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page summarizes key considerations related to regulatory expectations for biomanufacturing including chemicals and polymeric materials. For chemicals, topics include nitrosamines and elemental impurities, and for polymeric materials such as filters and single-use assemblies, the focus is qualification and extractable and leachable testing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service