Skip to Content
Merck
All Photos(1)

Key Documents

13-1490

Sigma-Aldrich

Hydrazinium dichloride

SAJ special grade, ≥98.0%

Synonym(s):

Hydrazine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · 2HCl
CAS Number:
Molecular Weight:
104.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

vapor pressure

5 hPa ( 20 °C)
7.69 hPa ( 25 °C)

Assay

≥98.0%

form

liquid

availability

available only in Japan

mp

200 °C (dec.) (lit.)

density

1.42 g/mL at 25 °C (lit.)

SMILES string

Cl[H].Cl[H].NN

InChI

1S/2ClH.H4N2/c;;1-2/h2*1H;1-2H2

InChI key

LIAWOTKNAVAKCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia A Mohamed et al.
Molecules (Basel, Switzerland), 17(12), 13969-13988 (2012-11-28)
Twelve novel intrinsically colored wholly aromatic azopolyamide-hydrazides containing various proportions of para- and meta-phenylene units were successfully synthesized by a low temperature (-10 °C) solution polycondensation reaction of either 4-amino-3-hydroxybenzhydrazide (4A3HBH) or 3-amino-4-hydroxybenzhydrazide (3A4HBH) with an equimolar amount of either
Weidong Qu et al.
Biosensors & bioelectronics, 42, 430-433 (2012-12-19)
Graphene-nickel nanoparticle hybrid was prepared by the one-step far infrared-assisted reduction of graphene oxide and nickel (II) ions using hydrazine. It was loaded on the surface of a magnetic electrode for electrochemical sensing. The feasibility and performance of the novel
Paul D Rainville et al.
Bioanalysis, 4(23), 2833-2842 (2012-12-12)
Accurate mass based LC-MS combined with statistical analysis is established as a core analytical technology for metabonomic studies. This is primarily due to the specificity, sensitivity and structural elucidation capabilities of the technology. The vast majority of these studies are
Hatem A Abdel-Aziz et al.
Molecules (Basel, Switzerland), 18(2), 2084-2095 (2013-02-08)
Although salicylaldehyde azine (3) was reported in 1985 as the single product of the reaction of ethyl 2-oxo-2H-chromene-3-carboxylate (1) with hydrazine hydrate, we identified another main reaction product, besides 3, which was identified as malono-hydrazide (4). In the last
Chirantan Kar et al.
Inorganic chemistry, 52(2), 743-752 (2013-01-11)
We have synthesized a new indole functionalized rhodamine derivative L(1) which specifically binds to Cu(2+) in the presence of large excess of other competing ions with visually observable changes in their electronic and fluorescence spectral behavior. These spectral changes are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service