Skip to Content
Merck
All Photos(1)

Key Documents

8.43816

Sigma-Aldrich

Hydrazinium dichloride

for synthesis

Synonym(s):

Hydrazinium dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€53.30
1 KG
€206.00

€53.30


Estimated to ship on16 April 2025Details



Select a Size

Change View
100 G
€53.30
1 KG
€206.00

About This Item

Empirical Formula (Hill Notation):
H4N2 · 2HCl
CAS Number:
Molecular Weight:
104.97
MDL number:
UNSPSC Code:
12352301
EC Index Number:
226-283-2
NACRES:
NA.22
solubility:
2700 g/L

€53.30


Estimated to ship on16 April 2025Details


Quality Level

potency

129 mg/kg LD50, oral (Rat)
91 mg/kg LD50, skin (Rabbit)

pH

0.5 (20 °C, 50 g/L in H2O)

mp

198 °C (decomposition)

solubility

2700 g/L

density

1.42 g/cm3 at 20 °C

bulk density

770 kg/m3

storage temp.

2-30°C

SMILES string

Cl.NN

InChI

1S/ClH.H4N2/c;1-2/h1H;1-2H2

InChI key

BIVUUOPIAYRCAP-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 97.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service