Skip to Content
Merck
All Photos(1)

Documents

03-3770

Sigma-Aldrich

Bromocresol Green

JIS special grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14Br4O5S
CAS Number:
Molecular Weight:
698.01
Beilstein:
372527
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

JIS special grade

availability

available only in Japan

mp

225 °C (dec.) (lit.)

SMILES string

Cc1c(Br)c(O)c(Br)cc1C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4C

InChI

1S/C21H14Br4O5S/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21/h3-8,26-27H,1-2H3

InChI key

FRPHFZCDPYBUAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margarida C Souza et al.
Talanta, 82(3), 1027-1032 (2010-08-04)
An automatic method for kinetics independent spectrometric analysis is proposed in this study. It uses a non-linear calibration model that explores concentration gradients generated by a flow-batch analyser (FBA) for the samples, dye, and the single standard solution. The procedure
Mehrorang Ghaedi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 62-68 (2011-11-23)
In the present research, cadmium hydroxide nanowire loaded on activated carbon (Cd(OH)(2)-NW-AC) was synthesized and characterized. This new adsorbent was applied for the removal of Bromocresol Green (BCG) molecules from aqueous solutions. The influence of effective variables such as solution
Philippe Meersseman et al.
The Journal of clinical endocrinology and metabolism, 100(7), 2613-2620 (2015-05-06)
Nutrition can affect the hypothalamus-pituitary-adrenal axis. We hypothesized that early administration of parenteral nutrition (PN) during critical illness reduces plasma ACTH and cortisol concentrations and thereby increases the use of corticosteroids. This is a preplanned substudy of a randomized controlled
Idris Guessous et al.
Clinical journal of the American Society of Nephrology : CJASN, 10(7), 1162-1169 (2015-04-23)
Molecular evidence suggests that levels of vitamin D are associated with kidney function loss. Still, population-based studies are limited and few have considered the potential confounding effect of baseline kidney function. This study evaluated the association of serum 25-hydroxyvitamin D
Kerstin Müller et al.
Veterinary clinical pathology, 39(1), 79-82 (2009-08-26)
In reptile medicine, plasma chemistry analysis is widely used for the evaluation of an individual's health status. The standard method for the determination of plasma albumin concentration is protein electrophoresis combined with the determination of total protein concentration, but the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service