Skip to Content
Merck
All Photos(1)

Key Documents

TBN30

Total Base Number Standard

ASTM D2896 (30.17 mg KOH/g), pkg of 1 × 50 g

Synonym(s):

TBN-Standard

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€287.00

€287.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€287.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€287.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

according to ASTM® D2896

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 × 50 g

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Total Acid Number Standards can be found on the Paragon Scientific webpage

Application

This CRM is intended for the calibration and validation of analytical instruments used to measure base number by potentiometric titration.

Features and Benefits

  • Manufactured and verified in an ISO 17025 and ISO 17034 dual accredited laboratory, as per UKAS
  • Fully traceable to recognized national standards, and to units of measurement recognized at the National Institute Standards and Technology (NIST) and other recognized national standards laboratories
  • Certified in accordance with ASTM D2896 and other equivalent procedures of IP 276, ISO 3771, and BS 2000 Part 276
  • Typically used in, but not limited to, the analysis of used oils and lubricants
  • Available in a single-volume pack of 125 g or as triple packs of the same size
  • Supplied in tamper-evident glass packaging, to assure sample integrity
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service