Skip to Content
Merck
All Photos(1)

Key Documents

11112589001

Roche

Blocking Reagent

For ELISA

Synonym(s):

blocking reagent

Sign Into View Organizational & Contract Pricing

Select a Size

27 G
€75.80

€75.80


Please contact Customer Service for Availability


Select a Size

Change View
27 G
€75.80

About This Item

UNSPSC Code:
12352200

€75.80


Please contact Customer Service for Availability

Quality Level

form

powder

packaging

pkg of 27 g

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Blocking reagent is used for blocking of nonspecific binding sites in ELISA procedures.

Application

For research use only. Not for use in diagnostic procedures.
Blocking reagent has been used as a blocking reagent in:
  • ELISA[1]
  • Immunofluorescence[2]
  • In situ hybridization[3]

Preparation Note

Storage conditions (working solution): Autoclaved stock solution can be stored for several days to one week either unopened at 15 to 25 °C or at 2 to 8 °C after opening. Alternatively, the stock solution can be stored in aliquots at -15 to -15 °C for up to 6 months.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Mravinacova et al.
New biotechnology, 66, 46-52 (2021-10-11)
Highly accurate serological tests are key to assessing the prevalence of SARS-CoV-2 antibodies and the level of immunity in the population. This is important to predict the current and future status of the pandemic. With the recent emergence of new
Anja K Wege et al.
mAbs, 6(4), 968-977 (2014-05-30)
Humanized tumor mice (HTM) were generated by the co-transplantation of human hematopoietic stem cells and human breast cancer cells overexpressing HER2 into neonatal NOD-scid IL2Rγ(null) (NSG) mice. These mice are characterized by the development of a human immune system in
David Drews et al.
BMC developmental biology, 9, 45-45 (2009-07-30)
The transcription factor Yin Yang 2 (YY2) shares a structural and functional highly homologue DNA-binding domain with the ubiquitously expressed YY1 protein, which has been implicated in regulating fundamental biological processes. However, the biological relevance of YY2 has not been
Laurens J Lambert et al.
PloS one, 8(12), e82110-e82110 (2013-12-11)
During B cell activation, the DNA lesions that initiate somatic hypermutation and class switch recombination are introduced by activation-induced cytidine deaminase (AID). AID is a highly mutagenic protein that is maintained in the cytoplasm at steady state, however AID is
Anneke H Hellinga et al.
International journal of molecular sciences, 21(23) (2020-12-03)
Bovine immunoglobulin G (bIgG) was previously shown to enhance innate immune responses to toll-like receptor (TLR) stimulation, via induction of trained immunity. In this study, we investigated whether minimally processed dairy streams with high levels of whey proteins as potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service