Skip to Content
Merck
All Photos(1)

Key Documents

20-200

Millipore

Immunoblot Blocking Reagent

20 g Immunoblot Blocking Reagent for Western Blotting.

Synonym(s):

Western Blot Blocking Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
€91.20

€91.20


Estimated to ship on11 April 2025Details



Select a Size

Change View
20 G
€91.20

About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

€91.20


Estimated to ship on11 April 2025Details


form

solid

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

shipped in

wet ice

Application

For use in western blots as a blocking agent.

Quality

Routinely evaluated by western blot

Physical form

non-fat dry milk fortified with vitamin A palmitate and vitamin D2, containing no preservatives

Storage and Stability

2 years at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florence Chalier et al.
Toxins, 12(2) (2020-02-09)
In the soft treatment of cancer tumours, consequent downregulation of the malignant tissue angiogenesis constitutes an efficient way to stifle tumour development and metastasis spreading. As angiogenesis requires integrin-promoting endothelial cell adhesion, migration, and vessel tube formation, integrins represent potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service