ZLXLSD52080
Milli-Q® HXSD Water Purification System
Centralized pure water solution for up to 1600 L/day of Type 2 water; for high chlorine feed water
Synonym(s):
Milli-Q® HX 7080 SD Water Purification System (HC)
About This Item
Recommended Products
AC/DC input
100 V / 230 V, 50-60 Hz
Quality Level
product line
Milli-Q® HX
packaging
pkg of 1 unit
parameter
1600 L/day max. usage
234 kg operating weight (515.9 lb)
80 L/hr make-up flow rate
resistivity
>5 MΩ-cm, at 25°C
system H × W × depth
124 cm (48.8 in.) × 54.3 cm (21.4 in.) × 94.7 cm (37.3 in.)
tank volume
140 L
total organic carbon (TOC) residue
<30 ppb (Product water)
input
potable tap water
output
type 2 water (> 5 MΩ·cm)
conductivity
<0.2 μS/cm
Related Categories
General description
Application
- Equipment and instruments such as dishwashers, autoclaves, clinical analyzers, heating baths, environmental and humidity chambers, and histopathology stainers.
- Taps on benches for general glassware rinsing, buffer and reagent preparation.
- Purification systems to obtain ultrapure water (e.g. Milli-Q® IQ 7000 system).
Features and Benefits
- Optimal combination of purification technologies for a reliable flow of high-quality Type 2 pure water.
- Integrated 140 L reservoir and pump for protected storage and distribution up to 20 L/min.
- Superior communications interface and modern data management capabilities.
- Customizable range of options and accessories to meet your specific requirements.
- MyMilli-Q™ Remote Care monitoring and service capability.
Components
- The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
- This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
- Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to
href="https://www.sigmaaldrich.com/campaigns/milli-q-iq7000-request-information?tfa_347=7011E000001KBVxQAO">contact us!
Other Notes
- Organism Retention: Microorganisms.
- Mode of Action: Filtering and UV.
- Application: General laboratory analysis.
- Intended Use: Water purification.
- Instructions for Use: This item provides water filtered through 0.22 ?m sterilizing-grade membranes and exposed to bactericidal UV lamp. Refer to system equipment user guide section Using the System (available on USB key in the system box).
- Storage Statement: Store in dry location.
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service