Skip to Content
Merck
All Photos(2)

Key Documents

ZAFS16D0WW

Milli-Q

AFS® Water Purification System

For analyzers with degassed pure water needs up to 320 L/day

Synonym(s):

AFS® 16D Water Purification System

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
Inquire

This item is not able to be purchased as a guest user. For further information, please contact us at 0800-14747.


Select a Size

Change View
1 EA
Inquire

About This Item

UNSPSC Code:
41104206
NACRES:
SB.53

This item is not able to be purchased as a guest user. For further information, please contact us at 0800-14747.

AC/DC input

100 V / 230 V, 50-60 Hz

Quality Level

product line

AFS®

packaging

pkg of 1 unit

parameter

0.6 L/min distribution flow rate (Dispensing to atmosphere (without any flow restriction due to the analyzer).)
16 L/hr make-up flow rate
22.2 kg operating weight (49 lb)
320 L/day max. usage

resistivity

>15 MΩ-cm, at 25 degrees

system H × W × depth

47 cm (18.5 in.) × 26.8 cm (10.6 in.) × 42.6 cm (16.8 in.)

total organic carbon (TOC) residue

<30 ppb

input

potable tap water

output

clinical laboratory reagent water (CLRW) (complies with the CLSI guideline*)

General description

AFS® D systems provide a cost-effective, low-maintenance water purification solution for analyzers with degassed pure water needs of up to 320 L/daily. The AFS® 16D water purification system has a production flow rate to the reservoir of 16 L/h and a dispensing flow rate to the analyzer of up to 0.6 L/min.
Water produced by AFS® systems complies with the Clinical and Laboratory Standards Institute (CLSI) guideline for clinical laboratory reagent water (CLRW). These systems combine complementary water purification techniques (complete pretreatment, advanced reverse osmosis (RO), bactericidal UV lamp, final 0.22 µm filtration) to provide reliable and consistent CLRW-quality water, with an additional step of degassing which prevents dissolved gases from interfering with the analysis.
AFS® D systems have full monitoring and water quality archiving capabilities that allows storage of up to six months of data.

Application

AFS® D water purification systems contain integrated degassing technology designed to provide a solution for applications requiring the removal of soluble and insoluble gases prior to analysis.
A hollow fiber degassing technique successfully prevents the formation of bubbles near optical sensors and in tubing, resulting in a fast, effective reduction of soluble gas.

Features and Benefits

  • Complementary water purification techniques coupled with a degassing technique provide consistent water quality meeting CLRW standards
  • Cost-effective thanks to optimized lifetimes for pretreatment and polishing packs and low feed water use
  • Fulfills accreditation needs (e.g. CAP 15189SM accreditation to the ISO 15189:2022 standard) with full monitoring and automatic water quality archiving capabilities. Up to 6 months of information can be stored for reliable traceability
  • An intuitive icon-based system display facilitates maintenance, and an ergonomic pack-locking system makes pack changes easier than ever
  • Milli-Q® Service Plans offer a range of support, including preventive maintenance visits and online contract management
  • A small footprint, allows installation wherever it’s convenient: on the wall, on or under the bench, or on a cart

Components

  • The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
  • This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore how high-quality water is essential for accurate clinical chemistry assays and complying with norms and guidelines (e.g. CLSI).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service