Skip to Content
Merck
All Photos(2)

Key Documents

ZAFS15EWW

Milli-Q

AFS® Water Purification System

Water purification system with Elix® technology, producing CLRW quality water up to 300 L/day

Synonym(s):

AFS® 15E Water Purification System

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
Inquire

This item is not able to be purchased as a guest user. For further information, please contact us at 0800-14747.


Select a Size

Change View
1 EACH
Inquire

About This Item

UNSPSC Code:
41104206
NACRES:
SB.53

This item is not able to be purchased as a guest user. For further information, please contact us at 0800-14747.

AC/DC input

100 - 230 V, 50 - 60 Hz

Quality Level

product line

AFS®

packaging

pkg of 1 unit

parameter

15 L/hr make-up flow rate
2 L/min distribution flow rate
2 L/min product water instant delivery rate
28 kg operating weight (61.7 lb)
300 L/day max. usage

resistivity

>15 MΩ-cm, at 25°C

system H × W × depth

58.5 cm (23 in.) × 26.8 cm (10.6 in.) × 42.6 cm (16.8 in.)

total organic carbon (TOC) residue

<30 ppb

input

potable tap water

output

clinical laboratory reagent water (CLRW) (that complies with the CLSI guideline)

Looking for similar products? Visit Product Comparison Guide

General description

AFS® E systems combine state-of-the-art Elix® electrodeionization (EDI) technology with other complementary water purification techniques to provide reliable and consistent water quality. The AFS® 15E water purification system can produce up to 300 L of water daily with a production flow rate to the reservoir of 15 L/h and a dispensing flow rate to the analyzer of 2 L/min. Water produced by the systems complies with the Clinical and Laboratory Standards Institute (CLSI) guideline for clinical laboratory reagent water (CLRW). Elix® EDI technology, which uses ion exchange resins that are permanently self-regenerated, helps ensure low and predictable running costs for the AFS® E range of systems. These systems have full monitoring and water quality archiving capabilities that allow storage of up to six months of data.

Application

AFS® E series water purification systems with Elix® technology, feed analyzers with consistent water quality that meets CLRW standards of the CLSI.

Features and Benefits

  • State-of-the-art Elix® electrodeionization (EDI) technology is combined with other complementary water purification techniques to provide consistent water quality meeting CLRW standards
  • Elix® EDI technology uses permanently self-regenerating ion exchange resins to ensure predictable running costs
  • Fulfills accreditation needs (e.g. CAP 15189SM accreditation to the ISO 15189:2022 standard) with full monitoring and automatic water quality archiving capabilities. Up to 6 months of information can be stored for reliable traceability
  • Patented Elix® technology provides constant quality water without the need for softeners or conditioning cartridges, which means reduced maintenance ― and less analyzer downtime
  • Milli-Q® Service Plans offer a range of support, including preventive maintenance visits and online contract management
  • A small footprint allows installation wherever it’s convenient: on the wall, on or under the bench, or on a cart

Components

  • The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
  • This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Other Notes

Directions For Use:
  • Organism Retention: Microorganisms
  • Mode of Action: Filtering and UV
  • Application: Clinical laboratories
  • Intended Use: Water purification
  • Instructions for Use: This item provides water filtered through 0.22 ?m sterilizing-grade membranes and exposed to bactericidal UV lamp. Please refer to system equipment user guide section "System" (provided on USB key in the North American configuration version).
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state, and local regulations.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Explore how high-quality water is essential for accurate clinical chemistry assays and complying with norms and guidelines (e.g. CLSI).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service